missing translation for 'onlineSavingsMsg'
Learn More

CD16/CD32, APC, clone: 93, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-0161-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15360580

  • £193.08 / 100µg


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 93 monoclonal antibody reacts with an epitope shared by mouse CD16 and CD32. CD16 (Fc gamma III Receptor) and CD32 (Fc gamma II Receptor) are the low affinity receptors for the mouse IgG Fc portion and are expressed by B cells, monocyte/macrophages, NK cells, and neutrophils.Applications Reported: The 93 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 93 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD16/CD32
93
APC
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
FCGR3, IGFR3; FCGR2, IGFR2; FC Receptor Block
IgG2a
RUO
Primary
Murine
Videos