Learn More
CD155 Monoclonal Antibody (2H7CD155), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 46-1550-42
202.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The monoclonal antibody 2H7CD155 recognizes CD155 also known as the poliovirus receptor. The CD155 protein is a member of the Ig superfamily in which 4 isoforms exist; only the alpha and delta forms are expressed on the membrane of monocytes, prostate cancer cells, as well as neurectodermal and monocytic tumor cells lines. In addition, CD155 is found during mid-gastrulation during embryonic development. CD155 is thought to play a role in adhesion by interaction with the ECM component vitronectin as well as a role in NK killing of tumor cells. CD226 and CD96 are ligands for CD155. Blocking of CD155 is under therapeutic investigation for prevention of variants of poliovirus. Applications Reported: This 2H7CD155 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 2H7CD155 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells or human tumor cell line U937. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative.
CD155 Monoclonal antibody specifically detects CD155 in Human samples. It is validated for Flow Cytometry.Specifications
CD155 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P15151 | |
PVR | |
Affinity chromatography | |
RUO | |
5817 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
2H7CD155 | |
PerCP-eFluor 710 | |
PVR | |
3830421F03Rik; AI325026; AI987993; CD112; CD155; D7Ertd458e; FLJ25946; herpes virus entry mediator B; Herpesvirus entry mediator B; hveB; HVED; mE4; mHveB; Mph; murine herpes virus entry protein B; murine herpesvirus entry protein B; NECL5; necl-5; Nectin cell adhesion molecule 2; Nectin2; Nectin-2; nectin-like 5; nectin-like protein 5; Poliovirus receptor; poliovirus receptor homolog; poliovirus receptor-related 2; poliovirus receptor-related protein 2; poliovirus sensitivity; PVR; Pvrl2; PVS; sCD112; soluble CD112; Taa1; TAGE4; tumor-associated antigen 1; tumor-associated glycoprotein pE4 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.