missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD154 (CD40 Ligand) Monoclonal Antibody (MR1), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen HMCD15401
Code : 50
Additional Details : Weight : 0.00010kg
Description
CD154 (CD40 Ligand) Monoclonal antibody specifically detects CD154 (CD40 Ligand) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD154 (CD40 Ligand) | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
P27548 | |
CD40LG | |
Mouse CD154. from Activated mouse Th1 clone D1.6 | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG3 |
Flow Cytometry | |
MR1 | |
FITC | |
CD40LG | |
CD154; CD40 antigen ligand; CD40 ligand; CD40 ligand CD154; CD40 ligand, membrane form; CD40 ligand, soluble form; CD40L; CD40-L; Cd40lg; gp39; hCD40L; H-CD-40-L; HIGM1; IGM; IMD3; Ly62; Ly-62; M-CD-40-L; RP23-153G22.3; T-B cell-activating molecule; T-BAM; T-cell antigen Gp39; TNF-related activation protein; TNFSF5; TRAP; tumor necrosis factor (ligand) superfamily member 5; tumor necrosis factor (ligand) superfamily, member 5; tumor necrosis factor ligand superfamily member 5 | |
Armenian Hamster | |
purified | |
RUO | |
21947 | |
4° C, store in dark | |
Liquid |