missing translation for 'onlineSavingsMsg'
Learn More

CD154 (CD40 Ligand) Mouse anti-Cynomolgus Monkey, Human, APC, Clone: 24-31, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1548-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15598016

  • £197.00 / 100 tests

In stock, ready to ship: 7

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD40 Ligand (CD40-L), or CD154, is a membrane glycoprotein and differentiation antigen expressed on the surface of T cells. The CD40 Ligand stimulates B cell proliferation and secretion of all immunoglobulin isotypes in the presence of cytokines. CD40 Ligand has been shown to induce cytokine production and tumoricidal activity in peripheral blood monocytes. It also co-stimulates proliferation of activated T cells and this is accompanied by the production of IFN-gamma, TNF-alpha, and IL-2.
Specifications

Specifications

CD154 (CD40 Ligand)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P29965
RP23-153G22.3, CD154, CD40-L, Cd40l, HIGM1, IGM, IMD3, Ly-62, Ly62, T-BAM, TRAP, Tnfsf5, gp39, T-cell antigen Gp39, TNF-related activation protein, tumor necrosis factor ligand superfamily member 5
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Cynomolgus Monkey, Human
Flow Cytometry
24-31
APC
CD40LG
Liquid
CD40LG
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
102138630, 959
Videos
SDS
Documents

For Research Use Only.