missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD153 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA568434
Additional Details : Weight : 0.01000kg
Description
CD153 Polyclonal antibody specifically detects CD153 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
CD153 | |
Polyclonal | |
Unconjugated | |
Tnfsf8 | |
CD153; CD153 antigen; CD30 antigen ligand; CD30 ligand; Cd30l; CD30-L; Cd30lg; CD30ligand; Tnfsf8; TNLG3A; tumor necrosis factor (ligand) superfamily member 8; tumor necrosis factor (ligand) superfamily, member 8; tumor necrosis factor ligand 3A; tumor necrosis factor ligand superfamily member 8; tumor necrosis factor superfamily member 8 | |
Rabbit | |
affinity chromatography | |
RUO | |
944 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P32971 | |
Tnfsf8 | |
Synthetic peptide corresponding to residues near the N terminal of human TNFSF8. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |