CD153 Rat anti-Mouse, Clone: RM153, eBioscience™

Rat Monoclonal Antibody

Overview
Brand: Affymetrix eBioscience

Manufacturer Part Number: 14-1531-82

100UG:ANTI-MOUSE CD153 PURIFIED 100 UG

UNSPSC: 12352200

Code: Z2

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only.

Product Code. 15217257

Quantity Price
1 £ 101.0 / 100µg
Estimated Shipment date
from Supplier 20-02-2017
Add to basket


Description and Specification

Specification

Antigen CD153
Format Purified
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Gene Alias CD30 Ligand, TNFSF8
Host Species Rat
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Isotype IgG2b
Quantity 100μg
Regulatory Status RUO
Species Reactivity Mouse
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone RM153
Concentration 0.5mg/mL
Conjugate Unlabeled

The RM153 monoclonal antibody reacts with mouse CD153 (CD30 ligand). This 40kDa cell surface receptor, a type II transmembrane protein, is a member of the TNF superfamily. Interaction of CD30 with CD153 plays an important role in T cell activation/costimulation. Expression of CD153 on T cells does not appear to require costimulation through CD28 and is upregulated via TCR engagement and peaks 24-48 hours after activation. Signaling through CD153 is important in costimulation of pre-activated T cells.