missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTLA-4 Monoclonal Antibody (1B8), FITC, Invitrogen™
Armenian Hamster Monoclonal Antibody
Brand: Invitrogen HMCD15201
241.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00010kg
Description
CTLA-4 Monoclonal antibody specifically detects CTLA-4 in Mouse samples. It is validated for ELISA, Flow Cytometry.Specifications
CTLA-4 | |
Monoclonal | |
0.5 mg/mL | |
PBS with <0.1% sodium azide | |
P09793 | |
CTLA4 | |
Mouse CD152 (CTLA-4). from Extracellular portion of murine CTLA-4 fused to a murine IgG2a | |
500 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry | |
1B8 | |
FITC | |
CTLA4 | |
ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form | |
Armenian Hamster | |
purified | |
RUO | |
12477 | |
4° C, store in dark | |
Liquid |