missing translation for 'onlineSavingsMsg'
Learn More

CD151 Mouse anti-Human, APC, Clone: 50-6, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1519-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15568016

  • £92.50 / 25 tests

Estimated Shipment date
from Supplier 06-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered. The protein encoded by this gene is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains. The proteins mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. This encoded protein is a cell surface glycoprotein that is known to complex with integrins and other transmembrane 4 superfamily proteins. It is involved in cellular processes including cell adhesion and may regulate integrin trafficking and/or function. This protein enhances cell motility, invasion and metastasis of cancer cells. Multiple alternatively spliced transcript variants that encode the same protein have been described for this gene.
Specifications

Specifications

CD151
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P48509
GP27, MER2, PETA-3, RAPH, SFA1, TSPAN24
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
50-6
APC
CD151
Liquid
CD151
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
977
Videos
Documents

For Research Use Only.