Learn More
CD150 Monoclonal Antibody (mShad150), PE-Cyanine7, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 25-1502-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The mShad150 monoclonal antibody reacts with mouse CD150, an ∽70 kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T cells, in particular Th1, and B cells; this expression is rapidly upregulated upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. The mShad150 antibody has been reported to also stain the CD150+CD244-CD48-CD41- population of pluripotent hematopoietic stem cells. Applications Reported: This mShad150 antibody has been reported for use in flow cytometric analysis. Applications Tested: This mShad150 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light.
CD150 Monoclonal antibody specifically detects CD150 in Mouse samples. It is validated for Flow Cytometry.Specifications
CD150 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q9QUM4 | |
Slamf1 | |
Affinity chromatography | |
RUO | |
27218 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
mShad150 | |
PE-Cyanine7 | |
Slamf1 | |
4933415F16; AA177906; CD150; CD150 antigen; CDw150; ESTM51; IPO-3; RGD1560634; signaling lymphocyte activation molecule; signaling lymphocytic activation molecule; signaling lymphocytic activation molecule family member 1; Slam; SLAM family member 1; Slamf1 | |
Rat | |
25 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b λ |
For Research Use Only.