missing translation for 'onlineSavingsMsg'
Learn More

CD150, eFluor 450, clone: mShad150, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 48-1502-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15529726

  • £195.81 / 100µg


Estimated Shipment date
from Supplier 28-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The mShad150 monoclonal antibody reacts with mouse CD150, an ∽70kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T cells, in particular Th1, and B cells; this expression is rapidly upregulated upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM in T cells induces proliferation and augmentation of the interferon-gamma response. Furthermore, SLAM is thought to play a role in adhesion between the T cell and antigen-presenting cell. The mShad150 antibody has been reported to also stain the CD150+CD244-CD48-CD41- population of pluripotent hematopoietic stem cells.Applications Reported: This mShad150 antibody has been reported for use in flow cytometric analysis.Applications Tested: This mShad150 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD150
mShad150
eFluor 450
Conjugated
Rat
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SLAM, Signaling lymphocyte activation molecule
IgG2b
RUO
Primary
Murine
Videos