missing translation for 'onlineSavingsMsg'
Learn More

CD150, FITC, clone: A12 (7D4), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1509-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15576496

  • £68.96 / 25 tests


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The A12 monoclonal antibody reacts with human CD150, an ∽70kDa transmembrane glycoprotein also known as Signaling Lymphocyte Activation Molecule (SLAM). CD150 is expressed by T and B cells and its expression is rapidly upregulated on these cells upon activation. Immature thymocytes and dendritic cells also express this antigen. Signaling through SLAM induces proliferation and cytokine production of T cells and SLAM plays a role in activation of T and B cells.Applications Reported: This A12 (7D4) antibody has been reported for use in flow cytometric analysis.Applications Tested: The A12 (7D4) antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (1 μg)/test per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488 nm; Emission: 520 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD150
A12 (7D4)
FITC
Conjugated
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (1μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
SLAM, IPO-3, IPO3, SLAMF1
IgG1
RUO
Primary
Human
Videos