Learn More
CD15 Monoclonal Antibody (HI98), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£300.00
Specifications
Antigen | CD15 |
---|---|
Clone | HI98 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15569086
|
Affymetrix eBioscience
46-0159-42 |
100 Tests |
£300.00
100 tests |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The HI98 monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative. Applications Reported: This HI98 antibody has been reported for use in flow cytometric analysis. Applications Tested: This HI98 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
CD15 Monoclonal antibody specifically detects CD15 in Human samples. It is validated for Flow Cytometry.Specifications
CD15 | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
stage-specific embryonic antigen 1; CD15; SSEA-1; SSEA1; ELFT; FCT3A; FUC-TIV; FUTIV; LeX | |
Primary | |
4° C, store in dark, DO NOT FREEZE! |
HI98 | |
Flow Cytometry | |
PerCP-eFluor 710 | |
Mouse | |
Human | |
IgM κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.