missing translation for 'onlineSavingsMsg'
Learn More

CD15, PE, clone: HI98, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0159-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15556716

  • £68.81 / 25 tests


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Description: The HI98 monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative.Applications Reported: This HI98 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HI98 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD15
Monoclonal
5μl/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
stage-specific embryonic antigen 1, CD15, SSEA-1, SSEA1, ELFT, FCT3A, FUC-TIV, FUTIV, LeX
IgM, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
Human
Flow Cytometry
HI98
PE
Conjugated
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Videos

For Research Use Only.