missing translation for 'onlineSavingsMsg'
Learn More

CD15 Mouse anti-Human, PE, Clone: HI98, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-0159-42

Code : NEW

Additional Details : Weight : 0.23750kg

Product Code. 15566716

  • £159.00 / 100 tests

In stock, ready to ship: 3

Please to purchase this item. Need a web account? Register with us today!


Descripción

Descripción

CD15 (Lewis X, Le(x); stage specific embryonic antigen-1, SSEA-1) is a trisacharide determit (3-fucosyl-N-acetyllactosamine) expressed on several glycolipids, glycoproteins and proteoglycans of various cell types, e.g. granulocytes, mast cells, monocytes, macrophages, cells of gastric mucosa, nervous system or various tumour cells. There are several variants of Lewis x, such as sialyl-Lewis x or sulphated Lewis x. Cells with high surface expression of Le(x) antigen exhibit strong self-aggregation, based on calcium-dependent Le(x)-Le(x) interaction. This process is involved for example in embryo compaction or in autoaggregation of teratocarcinoma cells. Sialyl-Le(x) and its isomer sialyl-Le(a) are ligands of selectins. CD15 expression has been extensively used to confirm diagnosis of Hodgkin´s disease.
Especificaciones

Especificaciones

CD15
Monoclonal
5 μL/Test
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2
stage-specific embryonic antigen 1; CD15; SSEA-1; SSEA1; ELFT; FCT3A; FUC-TIV; FUTIV; LeX
IgM, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
Human
Flow Cytometry
HI98
PE
Liquid
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Videos
missing translation for 'documents'

For Research Use Only.