Learn More
CD15 Monoclonal Antibody (HI98), eFluor™ 615, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 42-0159-80
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The HI98 monoclonal antibody reacts with human CD15, Lewis X. This 3-fucosyl-N-acetyllactosamine carbohydrate moiety is expressed by granulocytes. Monocytes express this structure at varying degree while lymphocytes are negative. Applications Reported: This HI98 antibody has been reported for use in immunohistochemical staing of frozen and FFPE tissue sections (IHC-F, IHC-P), and immunocytochemical staining. Applications Tested: This HI98 antibody has been tested by immunohistochemical staining of formalin-fixed paraffin embedded human human kidney cortex (IHC-P) using high pH antigen retrieval at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor™ 615 antibody conjugate, we recommend a filter that will capture the 615 emission wavelength (for example, Excitation 560/55, 585LP, Emission 645/75). A standard Alexa Fluor™ 594 filter is acceptable. Excitation: 595 nm; Emission: 615 nm. Filtration: 0.2 μm post-manufacturing filtered.
CD15 Monoclonal antibody specifically detects CD15 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
CD15 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P22083 | |
FUT4 | |
Affinity chromatography | |
RUO | |
2526 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
HI98 | |
eFluor 615 | |
FUT4 | |
stage-specific embryonic antigen 1; CD15; SSEA-1; SSEA1; ELFT; FCT3A; FUC-TIV; FUTIV; LeX | |
Mouse | |
25 μg | |
Primary | |
Human | |
Antibody | |
IgM κ |
For Research Use Only.