missing translation for 'onlineSavingsMsg'
Learn More

CD147, PE, clone: 8D12, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1472-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15556896

  • £62.63 / 25 tests


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The monoclonal antibody 8D12 recognizes CD147 which is a member of the immunoglobulin superfamily. It is also called EMMPRIN and basigin. CD147 has been characterized as a tumor cell collagenase stimulating molecule and has also been shown to have a role in cell adhesion. CD147 can be found in association with integrins alpha3 beta1 and alpha6 beta1 by co-immpunopreciptations. Additionally it is thought to play a role in T cell activation and thymocyte cycling. CD147 is expressed broadly on hematopoietic and non-hematopoietic cells, especially in tumor cells.Applications Reported: This 8D12 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 8D12 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD147
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P35613
EMMPRIN
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
8D12
PE
BSG
Conjugated
BSG
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
682
Videos

For Research Use Only.