missing translation for 'onlineSavingsMsg'
Learn More

CD147 Mouse anti-Human, PE, Clone: 8D12, eBioscience™

Mouse Monoclonal Antibody

£65.75 - £159.00

Specifications

Antigen CD147
Clone 8D12
Target Species Human
Conjugate PE
Isotype IgG1, kappa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15556896
View Documents
Affymetrix eBioscience
12-1472-41
25 Tests £65.75
25 tests View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Please to purchase this item. Need a web account? Register with us today!
15566896
View Documents
Affymetrix eBioscience
12-1472-42
100 Tests £159.00
100 tests View alternative products
Save Up to 
»
Estimated Shipment
27-11-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD147 (basigin, neurothelin, OX-47, 5A11, CE9, M6, EMMPRIN, extracellular matrix metalloproteinase inducer, TCSF, tumour cell-derived collagenase-stimulatory factor) is a 50-60 kDa glycoprotein with multiple glycosylated forms. CD147 is a plasma membrane protein that is important in spermatogenesis, embryo implantation, neural network formation, and tumor progression. CD147 is also a member of the immunoglobulin superfamily. The highest level of CD147 expression is on metabolically active cells, such as lymphoblasts, inflammatory cells, brown adipocytes and maligt tumor cells. CD147 has multiple functions, including facilitating of cell surface expression of monocarboxylate transporter proteins and extracellular matrix metalloproteinases, regulation of integrin functions, it plays roles in cell development and activation, fetal development or retinal function.
Specifications

Specifications

CD147
Human
IgG1, kappa
Monoclonal
Liquid
Antibody
4° C, store in dark, DO NOT FREEZE!
8D12
PE
Flow Cytometry
Primary
Monoclonal
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
RUO

For Research Use Only.