missing translation for 'onlineSavingsMsg'
Learn More

CD146 Mouse anti-Canine, Human, Mouse, Rabbit, FITC, Clone: P1H12, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1469-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15566496

  • £171.00 / 100 tests

EU Stock 2

Please to purchase this item. Need a web account? Register with us today!


Description

Description

2 μm post-manufacturing filtered. CD146 (MCAM) is an integral membrane glycoprotein belonging to the immunoglobulin superfamily. CD146 is expressed at cell-cell junctions in all endothelial cells, and has also been observed in other normal and maligt cell types. In addition to its recognized role as a calcium-independent cell adhesion molecule, CD146 may also act as a signal transduction molecule in the recruitment of Fyn kinase, and in the subsequent tyrosine-phosphorylation of intracellular proteins involved in actin cytoskeleton assembly. CD146 plays a role in cell adhesion, and in cohesion of the endothelial monolayer at intercellular junctions in vascular tissue. CD146 expression may allow melanoma cells to interact with cellular elements of the vascular system, thereby enhancing hematogeneous tumor spread. Further, CD146 could be an adhesion molecule active in neural crest cells during embryonic development. CD146 acts also as surface receptor that triggers tyrosine phosphorylation of FYN and PTK2, and a transient increase in the intracellular calcium concentration.
Specifications

Specifications

CD146
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Q8R2Y2, P43121
CD146; cell surface glycoprotein MUC18; cell surface glycoprotein P1H12; Gicerin; MCAM; melanoma adhesion molecule; melanoma-associated antigen A32; melanoma-associated antigen MUC18; MUC18; S-endo 1 endothelial-associated antigen
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Canine, Human, Mouse, Rabbit
Flow Cytometry
P1H12
FITC
MCAM
Liquid
Mcam
IgG1, kappa
100 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
100342848, 4162, 489368, 84004
Videos
Documents

For Research Use Only.