missing translation for 'onlineSavingsMsg'
Learn More

CD144 (VE-cadherin), Alexa Fluor 488, clone: eBioBV13 (BV13), eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 53-1441-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15342010

  • £125.34 / 25Āµg


Estimated Shipment date
from Supplier 26-07-2018
Add to basket

Description

Description

Description: The BV13 monoclonal antibody reacts with mouse VE-Cadherin (CD144). VE-Cadherin is a 120kDa member of the type II Cadherin family, characterized by the presence of 5 extracellular cadherin domains (ECD), and anchored to the actin cytoskeleton through their cytoplasmic tail. VE-Cadherin mediates homophilic adhesion between neighbouring endothelial cells and is localized within specialized structures at cell-cell contacts, called adherens junctions. VE-Cadherin is expressed constitutively throughout the entire vasculature, and is required for numerous endothelial cell functions including migration, survival, contact-dependent growth inhibition and endothelial cell assembly into tubular structures. Furthermore, it is thought that VE-Cadherin+CD45- cells from the yolk sac or aorta-gonad-mesonephros (AGM) have the potential to give rise to hematopoietic cells.Applications Reported: This eBioBV13 (BV13) antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections.Applications Tested: This eBioBV13 (BV13) antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD144
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P55284
Cadherin-5, Vascular endothelial cadherin, VE-cadherin, 7B4 antigen, CD144 antigen, CDH5
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow CytometryĆ¾Immunohistochemistry (Frozen)
eBioBV13 (BV13)
Alexa Fluor 488
Cdh5
Conjugated
Cdh5
IgG1, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
12562
Videos

For Research Use Only.