Learn More
CD144 (VE-cadherin) Monoclonal Antibody (eBioBV13 (BV13)), Alexa Fluor™ 488, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 53-1441-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The BV13 monoclonal antibody reacts with mouse VE-Cadherin (CD144). VE-Cadherin is a 120 kDa member of the type II Cadherin family, characterized by the presence of 5 extracellular cadherin domains (ECD), and anchored to the actin cytoskeleton through their cytoplasmic tail. VE-Cadherin mediates homophilic adhesion between neighbouring endothelial cells and is localized within specialized structures at cell-cell contacts, called adherens junctions. VE-Cadherin is expressed constitutively throughout the entire vasculature, and is required for numerous endothelial cell functions including migration, survival, contact-dependent growth inhibition and endothelial cell assembly into tubular structures. Furthermore, it is thought that VE-Cadherin+CD45- cells from the yolk sac or aorta-gonad-mesonephros (AGM) have the potential to give rise to hematopoietic cells. Applications Reported: This eBioBV13 (BV13) antibody has been reported for use in flow cytometric analysis, and immunohistology staining of frozen tissue sections. Applications Tested: This eBioBV13 (BV13) antibody has been tested by flow cytometric analysis of bEnd.3 cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD144 (VE-cadherin) Monoclonal antibody specifically detects CD144 (VE-cadherin) in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Frozen).Specifications
CD144 (VE-cadherin) | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P55284 | |
CDH5 | |
Affinity chromatography | |
RUO | |
12562 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry, Immunohistochemistry (Frozen) | |
eBioBV13 (BV13) | |
Alexa Fluor 488 | |
CDH5 | |
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG1 κ |
For Research Use Only.