Learn More
CD144 (VE-cadherin) Monoclonal Antibody (16B1), PE-Cyanine7, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 25-1449-41
64.66 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling. Applications Reported: This 16B1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 16B1 antibody has been pre-titrated and tested by flow cytometric analysis of HUVEC. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation.
CD144 (VE-cadherin) Monoclonal antibody specifically detects CD144 (VE-cadherin) in Human samples. It is validated for Flow Cytometry.Specifications
CD144 (VE-cadherin) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P33151 | |
CDH5 | |
Affinity chromatography | |
RUO | |
1003 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
16B1 | |
PE-Cyanine7 | |
CDH5 | |
7B4; 7B4 antigen; 7B4/cadherin-5; AA408225; cadherin 5; cadherin 5, type 2 (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular endothelium); cadherin 5, type 2, VE-cadherin (vascular epithelium); cadherin-5; cadherin-5; cd144 antigen; VE cadherin; Cd144; cd144 antigen; CDH5; cell-cell adhesion protein; endothelial-specific cadherin; vascular endothelial cadherin; vascular endothelial cadherin precursor; Vec; VEcad; VE-Cad; VE-cadherin; VECD | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 |
For Research Use Only.