missing translation for 'onlineSavingsMsg'
Learn More

CD144 (VE-cadherin), PE, clone: 16B1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1449-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15248679

  • £75.06 / 25µg


EU Stock 1
Add to basket

Description

Description

The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling.
Specifications

Specifications

CD144
16B1
PE
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Cadherin 5
IgG1
RUO
Primary
Human
Videos

For Research Use Only.