missing translation for 'onlineSavingsMsg'
Learn More

CD144 (VE-cadherin), clone: 16B1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-1449-80

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227237

  • £55.80 / 25µg


Estimated Shipment
27-02-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling.Applications Reported: This 16B1 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), microscopy, and immunocytochemistry without fixation.Applications Tested: This 16B1 antibody has been tested by immunoblotting Human Umbilical Vein Endothelial Cell (HUVEC) lysates. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD144 (VE-Cadherin)
16B1
Unconjugated
Purified
Murine
25μg
Store at 2-8°C.
Monoclonal
Western Blot
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
VECadherin, Cadherin 5, Cadherin5
IgG1
RUO
Primary
Human
Videos