missing translation for 'onlineSavingsMsg'
Learn More

CD144 (VE-cadherin), Biotin, clone: 16B1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1449-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299729

  • £62.63 / 25Āµg


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

Description: The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling.Applications Reported: This 16B1 antibody has been reported for use in flow cytometric analysis, and immunocytochemistry.Applications Tested: This 16B1 antibody has been tested by flow cytometric analysis of Human Umbilical Vein Endothelial Cells (HUVEC). This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD144 (VE-Cadherin)
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P33151
Cadherin-5, Vascular endothelial cadherin, VE-cadherin, 7B4 antigen, CD144 antigen, CDH5
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow CytometryĆ¾Immunocytochemistry
16B1
Biotin
CDH5
Conjugated
CDH5
IgG1
25μg
Store at 2-8°C. Do not freeze.
Primary
1003
Videos

For Research Use Only.