missing translation for 'onlineSavingsMsg'
Learn More

CD144 (VE-cadherin), Biotin, clone: 16B1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1449-82

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15209739

  • £137.47 / 100µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 16B1 antibody reacts with human CD144, also known as VE-cadherin and cadherin-5. The cadherin family of receptors, which are calcium-dependent adhesion molecules, is known to be involved in homophilic cell interactions. VE-cadherin, which is 140 kDa, is localized at the intercellular boundaries of endothelial cells in blood and lymphatic vessels in several tissues. It is thought to play a role in vascular permeability and remodeling.Applications Reported: This 16B1 antibody has been reported for use in flow cytometric analysis, and immunocytochemistry.Applications Tested: This 16B1 antibody has been tested by flow cytometric analysis of Human Umbilical Vein Endothelial Cells (HUVEC). This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD144 (VE-Cadherin)
16B1
Biotin
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze.
Monoclonal
Immunocytochemistry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
VECadherin, Cadherin 5, Cadherin5
IgG1
RUO
Primary
Human
Videos