missing translation for 'onlineSavingsMsg'
Learn More

CD142, APC, clone: HTF-1, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1429-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15598006

  • £62.12 / 25 tests


EU Stock 3
Add to basket

Description

Description

Description: This HTF-1 monoclonal antibody reacts with human CD142, which is also known as Tissue Factor. Expression of this type I transmembrane glycoprotein on endothelial cells, monocytes, macrophages, and platelets can be induced by inflammatory mediators (e.g., LPS, IL-1b, TNFa, PMA, or endotoxin). On the other hand, CD142 is expressed constitutively by some tumor cells (e.g., lung, pancreatic, breast, and colon) and non-immune tissues such as the vasculature, central nervous system, kidney, epithelia, and placenta. Studies have also suggested that CD142 exists as a soluble form that circulates in blood. CD142 initiates blood coagulation by associating with and activating the circulating factors VII and VIIa.The HTF-1 antibody has been reported to exhibit blocking activity.Applications Reported: This HTF-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: This HTF-1 antibody has been pre-titrated and tested by flow cytometric analysis of LPS-stimulated normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD142
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P13726
F3, Coagulation factor 3, TF, TFA
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
HTF-1
APC
F3
Conjugated
F3
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
2152
Videos

For Research Use Only.