missing translation for 'onlineSavingsMsg'
Learn More

CD141 Rat anti-Mouse, PE, Clone: LS17-9, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-1411-80

Code : NEW

Additional Details : Weight : 0.09500kg

Product Code. 15596886

  • £85.50 / 25µg

Estimated Shipment
27-11-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. Thrombomodulin (TM) is also known as Fetomodulin (FM), endothelial anticoagulant protein, or glycoprotein P112. Thrombomodulin contains six repeated domains homologous with epidermal growth factor (EGF) and an amino terminal domain homologous to lectin-like protein. Through its accelerated activation of protein C, synthesis of TM is one of several mechanisms important in maintaining thrombo-resistance and thus reducing clot formation on the surface of the endothelial cells. Immunohistochemical staining of formalin-fixed tissue for TM has been used for the study of a variety of vascular tumors, meningiomas and choriocarcinomas.
Specifications

Specifications

CD141
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P15306
AHUS6, BDCA3, CD141, THPH12, THRM, TM, CD141 antigen, fetomodulin, DC141, THBD
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
LS17-9
PE
Thbd
Liquid
Thbd
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
21824
Videos
Documents

For Research Use Only.