Learn More
CD140b (PDGFRB) Monoclonal Antibody (APB5), APC, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£92.25 - £240.00
Specifications
Antigen | CD140b (PDGFRB) |
---|---|
Clone | APB5 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15350680
|
Affymetrix eBioscience
17-1402-82 |
100 μg |
£240.00
100µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15340680
|
Affymetrix eBioscience
17-1402-80 |
25 μg |
£92.25
25µg |
Estimated Shipment: 30-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Description: The APB5 monoclonal antibody reacts with the mouse CD140b molecule, the beta chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRb is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of alpha/alpha, beta/beta, or heterodimers of alpha/beta and serve as a substrate for its kinase activity. CD140b is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues. The PDGFR beta chain is reported to play a significant role in formation of fibrous atherosclerotic lesions. Applications Reported: This APB5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This APB5 antibody has been tested by flow cytometric analysis of NIH 3T3 cell line. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
CD140b (PDGFRB) Monoclonal antibody specifically detects CD140b (PDGFRB) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD140b (PDGFRB) | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
AI528809; Beta platelet-derived growth factor receptor; beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140B; CD140b antigen; IBGC4; IMF1; JTK12; KOGS; PDGF beta chain; PDGF Receptor beta; Pdgfr; Pdgfr1; PDGFR-1; PDGFRB; PDGF-R-beta; PDGFR-beta; PENTT; platelet derived growth factor receptor beta; platelet derived growth factor receptor, beta polypeptide; platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; Platelet-derived growth factor receptor, beta; platelet-derived growth factor receptor, beta polypeptide | |
Pdgfrb | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
Pdgfrb |
APB5 | |
Flow Cytometry | |
APC | |
Rat | |
Mouse | |
P05622 | |
18596 | |
IgG2a κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.