missing translation for 'onlineSavingsMsg'
Learn More

CD140a (PDGFRA), PE-Cyanine7, clone: APA5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-1401-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15518486

  • £183.67 / 100µg


EU Stock 5
Add to basket

Description

Description

Description: The APA5 monoclonal antibody reacts with the mouse CD140a molecule, the alpha chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRa is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of alpha/alpha, beta/beta, or heterodimers of alpha/beta and serve as a substrate for its kinase activity. CD140a is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues.Applications Reported: This APA5 antibody has been reported for use in flow cytometric analysis.Applications Tested: This APA5 antibody has been tested by flow cytometric analysis of NIH/3T3 cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD140a (PDGF Receptor a)
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P26618
PDGF Receptor alpha, MGC74795, PDGFR2, RHEPDGFRA, CD140 antigen-like family member A, CD140a antigen, PDGF-R-alpha, PDGFRA/BCR fusion, pdgfr-a
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
APA5
PE-Cyanine7
Pdgfra
Conjugated
Pdgfra
IgG2a, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage, handling and experimental procedures.
Primary
18595
Videos

For Research Use Only.