missing translation for 'onlineSavingsMsg'
Learn More

CD140a (PDGFRA), clone: APA5, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 14-1401-81

Code : Z2

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15227227

  • £36.08 / 50µg


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Description: The APA5 monoclonal antibody reacts with the mouse CD140a molecule, the alpha chain of the platelet derived growth factor receptor (PDGF receptor). PDGFRa is a receptor tyrosine kinase that forms dimers on the surface upon ligand binding and phosphorylates substrates. Dimers of PDGFR consist of either homodimers of alpha/alpha, beta/beta, or heterodimers of alpha/beta and serve as a substrate for its kinase activity. CD140a is expressed by embryonic tissues and mesenchymal-derived cells of the adult mouse tissues.Applications Reported: The APA5 antibody has been reported for use in flow cytometric analysis, immunoblotting (WB), and immunohistochemical staining of frozen tissue sections. It has also been reported in blocking of ligand binding. (Please use Functional Grade purified APA5 in functional assays.).Applications Tested: This APA5 antibody has been tested by flow cytometric analysis of the NIH/3T3 cell line. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD140a
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P26618
PDGF Receptor alpha, MGC74795, PDGFR2, RHEPDGFRA, CD140 antigen-like family member A, CD140a antigen, PDGF-R-alpha, PDGFRA/BCR fusion, pdgfr-a
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Western Blot
APA5
Unconjugated
Pdgfra
Purified
Pdgfra
IgG2a, kappa
50μg
Store at 2-8°C.
Primary
18595
Videos

For Research Use Only.