missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ CD14 Recombinant Human Protein, N-His tag

Expressed from DNA sequence encoding mature form of human CD14 (Met 1-Met 344) fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 10073H08H5

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15204605

  • £56.21 / 5µg


Estimated Shipment
11-06-2018
Add to basket

Description

Description

  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein structure
929
≥90% by SDS-PAGE
Human
Truncated, Recombinant
Cluster of Differentiation 14 (CD14)
Assay Standard
Mammalian
N-His Tag
Lyophilized
36.5kd
5μg
−20°C
<1 EU/μg
CD14
CD Proteins
CD14 Recombinant Human Protein
Videos

For Research Use Only. Not for use in diagnostic procedures.