Learn More
Sino Biological™ CD14 Recombinant Human Protein, N-His tag
Expressed from DNA sequence encoding mature form of human CD14 (Met 1-Met 344) fused to polyhistidine tag at C-terminus
Brand: Sino Biological™ 10073H08H50
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Expressed from DNA sequence encoding mature form of human CD14 (Met 1-Met 344) fused to polyhistidine tag at C-terminus
- Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications
Assay Standard | |
36.5kd | |
Human | |
Truncated, Recombinant | |
Cluster of Differentiation 14 (CD14) | |
CD14 Recombinant Human Protein | |
Mammalian | |
Lyophilized |
929 | |
50μg | |
−20°C | |
<1 EU/μg | |
CD14 | |
N-His Tag | |
CD Proteins | |
≥90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.