missing translation for 'onlineSavingsMsg'
Learn More

CD14, Biotin, clone: Sa2-8, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0141-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15259459

  • £49.60 / 50µg


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The Sa2-8 monoclonal antibody reacts with mouse CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is a receptor for the complexes of LPS and LBP (LPS-Binding Protein) and is shown to associate with Toll-Like Receptor 4 (TLR4) and participate in the signaling and cellular response to bacterial LPS. In mouse, CD14 is expressed on the surface of macrophages and under certain conditions is also found in the serum in a secreted form. Sa2-8 has weak antagonistic activity (in NF-kappaB activation or TNF alpha production with LPS stimulation).Applications Reported: The Sa2-8 antibody has been reported for use in flow cytometric analysis.Applications Tested: This Sa2-8 antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal exudate cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD14
Sa2-8
Biotin
Conjugated
IgG2a
RUO
Primary
Murine
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Rat
50μg
Store at 2-8°C. Do not freeze.
Monoclonal
Videos