Learn More
CD14 Monoclonal Antibody (Sa2-8), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 13-0141-82
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The Sa2-8 monoclonal antibody reacts with mouse CD14, a 53-55 kDa GPI-linked glycoprotein. CD14 is a receptor for the complexes of LPS and LBP (LPS-Binding Protein) and is shown to associate with Toll-Like Receptor 4 (TLR4) and participate in the signaling and cellular response to bacterial LPS. In mouse, CD14 is expressed on the surface of macrophages and under certain conditions is also found in the serum in a secreted form. Sa2-8 has weak antagonistic activity (in NF-kappaB activation or TNF alpha production with LPS stimulation). Applications Reported: The Sa2-8 antibody has been reported for use in flow cytometric analysis. Applications Tested: This Sa2-8 antibody has been tested by flow cytometric analysis of mouse thioglycolate-elicited peritoneal exudate cells. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.Specifications
CD14 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
P10810 | |
Cd14 | |
Affinity chromatography | |
RUO | |
12475 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
Sa2-8 | |
Biotin | |
Cd14 | |
CD 14; CD14; CD14 antigen; CD14 molecule; cd14 monocyte; lipopolysaccharide receptor; lipoprotein receptor; LPSR antibody; monocyte differentiation antigen CD14; Monocyte differentiation antigen CD14, membrane-bound form; Monocyte differentiation antigen CD14, urinary form; monocyte differentiation antigen CD14; LOW QUALITY PROTEIN: monocyte differentiation antigen CD14; myeloid cell-specific leucine-rich glycoprotein; myeloid membrane glycoprotein precursor; sCD14; soluble CD14 | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.