missing translation for 'onlineSavingsMsg'
Learn More

CD14, eFluor 506, clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 69-0149-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510657

  • £68.86 / 25 tests


EU Stock 3
Add to basket

Description

Description

Description: The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.Applications Reported: This 61D3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 61D3 antibody has been pre-titrated and tested by flow cytometric analysis of normal peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 506 can be excited with the violet laser line (405 nm) and emits at 506 nm. We recommend using a 510/20 band pass filter, or equivalent. Please make sure that your instrument is capable of detecting this fluorochrome.Excitation: 405 nm; Emission: 506 nm; Laser: Violet Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD14
Monoclonal
5μl/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08571
monocyte differentiation antigen CD14, myeloid cell-specific leucine-rich glycoprotein
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
61D3
eFluor 506
CD14
Conjugated
CD14
IgG1, kappa
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
929
Videos

For Research Use Only.