missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, eVolve 605, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 83-0149-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15520747

  • £97.22 / 25 tests


Call for latest availability
+44 (0)1509 555 500
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.
Specifications

Specifications

CD14
61D3
eVolve 605
Conjugated
IgG1
RUO
Primary
Human
Flow Cytometry
5μL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This product should be used within 6 months from the date of receipt.
Monoclonal