missing translation for 'onlineSavingsMsg'
Learn More

CD14, Biotin, clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0149-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15289459

  • £56.04 / 25µg


Estimated Shipment date
from Supplier 29-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.Applications Reported: This 61D3 antibody has been reported for use in flow cytometric analysis.Applications Tested: This 61D3 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD14
61D3
Biotin
Conjugated
Murine
25μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LPS-R, Mo2
IgG1
RUO
Primary
Human
Videos