missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, Biotin, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-0149-82

UNSPSC : 12352200

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299459

  • £115.56 / 100µg


EU Stock 22
Add to basket

Disclaimers:

For Research Use Only.


Specifications

Description

The 61D3 monoclonal antibody reacts with human CD14, a 53-55kDa GPI-linked glycoprotein. CD14 is expressed on monocytes, interfollicular macrophages and some dendritic cells. Complexes of LPS and LBP (LPS-Binding Protein) bind with high affinity to monocytes through the surface CD14.
Description & Specifications

Specifications

CD14
61D3
Biotin
Conjugated
Murine
100μg
Store at 2-8°C. Do not freeze.
Monoclonal
Flow Cytometry
0.5mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
LPS-R, Mo2
IgG1
RUO
Primary
Human