missing translation for 'onlineSavingsMsg'
Learn More

CD14 Mouse anti-Human, Alexa Fluor® 700, Clone: 61D3, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 56-0149-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15580337

  • £70.00 / 25 tests

Estimated Shipment date
from Supplier 11-05-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

CD14 is a 55 kDa GPI-anchored glycoprotein that is constitutively expressed on the surface of mature monocytes, macrophages, and neutrophils. CD14 also serves as a multifunctional lipopolysaccharide receptor, and is released to the serum both as a secreted and enzymatically cleaved GPI-anchored form. CD14 binds lipopolysaccharide molecule in a reaction catalyzed by lipopolysaccharide-binding protein (LBP), an acute phase serum protein. The soluble sCD14 can discriminate slight structural differences between lipopolysaccharides and is important for neutralization of serum allochthonous lipopolysaccharides by reconstituted lipoprotein particles. Further, CD14 has been shown to bind apoptotic cells, and can affect allergic, inflammatory and infectious processes. Alternative splicing results in multiple transcript variants encoding the same CD14 isoform. Diseases associated with CD14 dysfunction include mycobacterium chelonae infection and Croup.
Specifications

Specifications

CD14
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P08571
monocyte differentiation antigen CD14, myeloid cell-specific leucine-rich glycoprotein
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
61D3
Alexa Fluor™ 700
CD14
Liquid
CD14
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
929
Videos
Documents

For Research Use Only.