missing translation for 'onlineSavingsMsg'
Learn More

CD138 (Syndecan-1), PE-Cyanine7, clone: DL-101, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 25-1389-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15588476

  • £113.58 / 25 tests


Estimated Shipment date
from Supplier 26-03-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The DL-101 monoclonal antibody reacts with human CD138, also known as syndecan-1. CD138 is a transmembrane protein containing chondroitin sulfate and heparin sulfate moieties responsible for binding to the extracellular matrix components. CD138 is not expressed by mature B cells but is present on pre-B cells and the finally differentiated plasma cells.Applications Reported: This DL-101 antibody has been reported for use in flow cytometric analysis.Applications Tested: This DL-101 antibody has been pre-titrated and tested by flow cytometric analysis of the U266 cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD138 (Syndecan-1)
DL-101
PE-Cyanine7
Conjugated
IgG1
RUO
Primary
Human
Flow Cytometry
5μL (0.5μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Murine
25 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Monoclonal
Videos