missing translation for 'onlineSavingsMsg'
Learn More

CD137 Polyclonal Antibody, PeproTech®, Invitrogen™

Goat Polyclonal Antibody

Brand:  Invitrogen 500-P167G-1MG

 View more versions of this product

Product Code. 17804083

  • £1374.00 / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MERTRSLQDP CSNCPAGTFC DNNRNQICSP CPPNSFSSAG GQRTCDICRQ CKGVFRTRKE CSSTSNAECD CTPGFHCLGA GCSMCEQDCK QGQELTKKGC KNCCFGTFND QKRGICRPWT NCSLDGKSVL VNGTKERDVV CGPSPADLSP GASSVTPPAP AREPGHS. Preparation: Produced from sera of goats immunized with highly pure Recombinant Human 4-1BB Receptor. Anti-Human 4-1BB Receptor-specific antibody was purified by affinity chromatography employing an immobilized Human 4-1BB Receptor matrix. Sandwich ELISA: To detect Human 4-1BB Receptor by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human 4-1BB Receptor (500-P167GBt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human 4-1BB Receptor. Western Blot: To detect Human 4-1BB Receptor by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Human 4-1BB Receptor is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD137
Polyclonal
Unconjugated
TNFRSF9
4 1 BB; 41BB; 4-1BB; 4-1BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1BB; s4 1 BB; s4 1BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1BB; soluble 41BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1BB; T-cell antigen 4-1BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9
Goat
Antigen affinity chromatography
RUO
3604
-20°C
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
Q07011
TNFRSF9
E.coli-derived Recombinant Human 4-1BB Receptor
1 mg
Primary
Human
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers