missing translation for 'onlineSavingsMsg'
Learn More

CD137 Ligand (4-1BB Ligand), PE, clone: TKS-1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5901-83

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15298899

  • £189.15 / 200µg


Estimated Shipment date
from Supplier 03-04-2018
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The TKS-1 monoclonal antibody reacts with mouse 4-1BB Ligand (4-1BBL), a type II transmembrane protein and a member of the tumor necrosis factor superfamily. 4-1BBL is expressed by activated antigen presenting cells including B cells and macrophages. Interaction of 4-1BBL with 4-1BB (CDw137) is reported to deliver a costimulatory signal important in the T-APC interaction.Applications Reported: The TKS-1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The TKS-1 antibody has been tested by flow cytometric analysis of resting and 2-3 day (IgM+CD40)-activated mouse splenocytes. This can be used at less than or equal to 1μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD137 Ligand
TKS-1
PE
Conjugated
Rat
200μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
4-1BBL, TNFSF9
IgG2a
RUO
Primary
Murine
Videos