missing translation for 'onlineSavingsMsg'
Learn More

CD135 (Flt3), PerCP-eFluor™ 710, clone: A2F10, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 46-1351-80

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15301290

  • £64.90 / 25µg


EU Stock 9
Add to basket

Description

Description

Description: The A2F10 monoclonal antibody reacts with mouse CD135, also known as Flk2/Flt3. A member of the tyrosine kinase receptor family, this 135-150kDa molecule is expressed by primitive progenitor cells in fetal liver and adult bone marrow. Two-color staining of adult mouse bone marrow cells with A2F10 and RA3-6B2 (CD45R/B220) or M1/70 (CD11b) reveals Flk-2-positive subpopulations of B cells or myeloid cells, respectively.Applications Reported: This A2F10 antibody has been reported for use in flow cytometric analysis.Applications Tested: This A2F10 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.PerCP-eFluor® 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome.Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically.Excitation: 488 nm; Emission: 710 nm; Laser: Blue Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD135
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q00342
RP11-153M24.3, CD135, FLK2, STK1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
A2F10
PerCP-eFluor 710
Flt3
Conjugated
Flt3
IgG2a, kappa
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
14255
Videos

For Research Use Only.