Learn More
CD135 (Flt3) Monoclonal Antibody (A2F10), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
Brand: Affymetrix eBioscience 46-1351-82
161.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : Z2
Additional Details : Weight : 0.25000kg
Description
Description: The A2F10 monoclonal antibody reacts with mouse CD135, also known as Flk2/Flt3. A member of the tyrosine kinase receptor family, this 135-150 kDa molecule is expressed by primitive progenitor cells in fetal liver and adult bone marrow. Two-color staining of adult mouse bone marrow cells with A2F10 and RA3-6B2 (CD45R/B220) or M1/70 (CD11b) reveals Flk-2-positive subpopulations of B cells or myeloid cells, respectively. Applications Reported: This A2F10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This A2F10 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. PerCP-eFluor™ 710 emits at 710 nm and is excited with the blue laser (488 nm); it can be used in place of PerCP-Cyanine5.5. We recommend using a 710/50 bandpass filter, however, the 695/40 bandpass filter is an acceptable alternative. Please make sure that your instrument is capable of detecting this fluorochrome. Fixation: Samples can be stored in IC Fixation Buffer (cat.
CD135 (Flt3) Monoclonal antibody specifically detects CD135 (Flt3) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD135 (Flt3) | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Q00342 | |
FLT3 | |
Affinity chromatography | |
RUO | |
14255 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
A2F10 | |
PerCP-eFluor 710 | |
FLT3 | |
B230315G04; CD135; CD135 antigen; fetal liver kinase 2; Fetal liver kinase-2; FL cytokine receptor; FLK2; Flk-2; FLT3; Flt-3; fms related tyrosine kinase 3; FMS-like tyrosine kinase 3; fms-related tyrosine kinase 3; growth factor receptor tyrosine kinase type III; Ly72; OTTHUMP00000214829; Receptor-type tyrosine-protein kinase FLT3; RP11-153M24.3; stem cell tyrosine kinase 1; STK1; STK-1; tyrosine-protein kinase FLT3; tyrosine-protein kinase receptor flk-2; wmfl | |
Rat | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
For Research Use Only.