missing translation for 'onlineSavingsMsg'
Learn More

CD135 (Flt3), Biotin, clone: A2F10, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 13-1351-81

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15299719

  • £50.12 / 50µg


Estimated Shipment date
from Supplier 04-06-2018
Add to basket

Description

Description

Description: The A2F10 monoclonal antibody reacts with mouse CD135, also known as Flk2/Flt3. A member of the tyrosine kinase receptor family, this 135-150kDa molecule is expressed by primitive progenitor cells in fetal liver and adult bone marrow. Two-color staining of adult mouse bone marrow cells with A2F10 and RA3-6B2 (CD45R/B220) or M1/70 (CD11b) reveals Flk-2-positive subpopulations of B cells or myeloid cells, respectively.Applications Reported: The A2F10 antibody has been reported for use in flow cytometric analysis.Applications Tested: The A2F10 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This can be used at less than or equal to 1 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD135
Monoclonal
0.5mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Q00342
RP11-153M24.3, CD135, FLK2, STK1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Flow Cytometry
A2F10
Biotin
Flt3
Conjugated
Flt3
IgG2a, kappa
50μg
Store at 2-8°C. Do not freeze.
Primary
14255
Videos

For Research Use Only.