missing translation for 'onlineSavingsMsg'
Learn More

CD134 (OX40) Mouse anti-Human, FITC, Clone: ACT35 (ACT-35), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 11-1347-41

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15596486

  • £66.00 / 25 tests

Estimated Shipment
01-12-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The protein encoded by this gene is a member of the TNF-receptor superfamily. This receptor has been shown to activate NF-kappaB through its interaction with adaptor proteins TRAF2 and TRAF5. Knockout studies in mice suggested that this receptor promotes the expression of apoptosis inhibitors BCL2 and BCL2lL1/BCL2-XL, and thus suppresses apoptosis. The knockout studies also suggested the roles of this receptor in CD4+ T cell response, as well as in T cell-dependent B cell proliferation and differentiation.
Specifications

Specifications

CD134 (OX40)
Monoclonal
5 μL/Test
PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
P43489
cluster designation 134, TNFRSF4
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
ACT35 (ACT-35)
FITC
TNFRSF4
Liquid
TNFRSF4
IgG1, kappa
25 Tests
4° C, store in dark, DO NOT FREEZE!
Primary
7293
Videos
Documents

For Research Use Only.