missing translation for 'onlineSavingsMsg'
Learn More

CD134 (OX40), APC, clone: ACT35 (ACT-35), eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 17-1347-42

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15528006

  • £149.92 / 100 tests


EU Stock 5
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Description: The ACT35 monoclonal antibody reacts with human CD134, also known as OX-40. A member of the TNF receptor superfamily, CD134/OX-40 is a 50kDa type I membrane glycoprotein expressed by activated T lymphocytes. The interaction of CD134 with OX-40L has been implicated in T cell-dependent humoral response, regulation of primary T cell expansion, survival of T cells, size of the memory T cell pool and regulation of tolerance in the CD4^+ T cell compartment.Applications Reported: The ACT35 (ACT-35) antibody has been reported for use in flow cytometric analysis.Applications Tested: This ACT35 (ACT-35) antibody has been pre-titrated and tested by flow cytometric analysis of 3-day PHA-stimulated normal human peripheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

CD134
ACT35 (ACT-35)
APC
Conjugated
Murine
100 Tests
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
5μL (0.125μg)/test
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
OX-40, TNFRSF4
IgG1
RUO
Primary
Human
Videos