Learn More
CD133 (Prominin-1) Monoclonal Antibody (EMK08), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 17-1339-41
70.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
The CD133 (also named AC133) antigen is a 97kDa glycoprotein, with 5 transmembrane domains, that shows up as a 120kDa band on protein gels due to glycosylation. It is a cell surface marker expressed by immature hematopoietic stem cells but not mature blood cells. In addition to hematopoietic stem and progenitor cells, CD133 has been found to be a marker for other stem and progenitor cells including neural and embryonic stem cells. It has been shown to be expressed in cancers, including some leukemias and brain tumours, that may be derived from stem cells. Low level expression could also be detected in the kidney, pancreas, placenta, and fetal liver.
CD133 (Prominin-1) Monoclonal antibody specifically detects CD133 (Prominin-1) in Human samples. It is validated for Flow Cytometry.Specifications
CD133 (Prominin-1) | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
O43490 | |
PROM1 | |
Affinity chromatography | |
RUO | |
8842 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
EMK08 | |
APC | |
PROM1 | |
4932416E19Rik; AC133; Antigen AC133; antigen AC133 homolog; antigen CD133; CD107b; CD133; CORD12; fudenine; hematopoietic stem cell antigen; hProminin; Lamp II; Lamp2; LAMP-2; Lamp-2a; Lamp-2b; Lamp-2c; LGP-96; LGP-B; Mac3; MCDR2; MSTP061; Prom; PROM1; Prom-1; prominin 1; prominin 1.s1; Prominin1; prominin-1; prominin-1.s2; prominin-like 1; prominin-like protein 1; PROML1; RP41; STGD4 | |
Mouse | |
25 Tests | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
For Research Use Only.