Learn More
CD132 Monoclonal Antibody (TUGh4), Biotin, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£72.25 - £167.00
Specifications
Antigen | CD132 |
---|---|
Clone | TUGh4 |
Concentration | 0.5 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15269719
|
Affymetrix eBioscience
13-1329-82 |
100 μg |
£167.00
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15259719
|
Affymetrix eBioscience
13-1329-80 |
25 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Description: This TUGh4 monoclonal antibody reacts with human CD132, the common gamma chain (gamma c) shared by the IL-2, IL-4, IL-7, IL-9, and IL-15 receptors. This approximately 65 kDa receptor component is expressed on T and B lymphocytes, NK cells, monocytes, and granulocytes. CD132 is involved in intracellular signal transduction by associating with effector molecules within the cytoplasm. Mutations in the human gamma chain gene result in X-linked severe combined immunodeficiency (XSCID), a disease characterized by T cell deficiency leading to impairment of cell-mediated and humoral immunity. Applications Reported: This antibody has been reported for use in flow cytometric analysis. Applications Tested: This TUGh4 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 μm post-manufacturing filtered.
IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.Specifications
CD132 | |
0.5 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
P31785 | |
IL2RG | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
IL2RG |
TUGh4 | |
Flow Cytometry | |
Biotin | |
Rat | |
Human | |
3561 | |
IgG2b λ | |
Affinity chromatography | |
Antibody |
For Research Use Only.