Learn More
CD131 Monoclonal Antibody (1C1), PE, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 12-1319-42
110.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.23750kg
Description
Description: The 1C1 monoclonal antibody reacts with the human CD131 molecule, also known as the common beta subunit (betaC). The common beta subunit associates with the specific alpha subunits of IL-3 receptor, IL-5 receptor and GM-CSF receptor to form high affinity receptors for these cytokines. These cytokine receptors are expressed by neutrophils, eosinophils, monocytes, endothelial cells, fibroblasts and hematopoietic progenitor cells and play a crucial role in growth/activation of eosinophils and in the inflammatory response. Applications Reported: This 1C1 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1C1 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
The protein encoded by this gene is the common beta chain of the high affinity receptor for IL-3, IL-5 and CSF. Defects in this gene have been reported to be associated with protein alveolar proteinosis (PAP).Specifications
CD131 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P32927 | |
Csf2rb | |
Affinity chromatography | |
RUO | |
1439 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
1C1 | |
PE | |
Csf2rb | |
Ai2ca; AI848964; AIC2A; Aic2b; Bc; beta c; beta common cytokine receptor; BetaIl3; Bil3; CD antigen CD131; CD131; CD131 antigen; CDw131; colony stimulating factor 2 receptor beta common subunit; colony stimulating factor 2 receptor, beta 1, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta 2, low-affinity (granulocyte-macrophage); colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage); colony-stimulating factor 2 receptor subunit beta-2; colony-stimulating factor-2 receptor, beta, low-affinity; common beta chain; CSF2RB; Csf2rb1; Csf2rb2; Csfgmrb; cytokine receptor common subunit beta; GM-CSF/IL-3/IL-5 receptor common beta subunit; GM-CSF/IL-3/IL-5 receptor common beta-chain; IL-3 RB; IL-3 Receptor beta; IL-3 receptor class 2 subunit beta; Il3r; IL3R beta; IL-3R class 2 subunit beta; IL3RB; Il3rb1; Il3rb2; IL5RB; interleukin 3 common beta chain; interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3 receptor, | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
For Research Use Only.